2,3,5-Trimethylphenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
T78603-100G
T78603-5G
SIALT78603-100GEA
76.3
GBP
SIALT78603-100G
SIALT78603-5G
2,3,5-Trimethylphenol, Sigma-Aldrich®
2,3,5-Trimethylphenol
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 230 °C (1013 hPa) Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002228 CAS Number: 697-82-5 EINECS: 211-806-9 UN: 2430 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...