2,4,6-Trimethylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Isothiocyanato-2,4,6-trimethylbenzene, 2,4,6-Trimethylphenylisothiocyanate

009586-5G 009586-25G
FLUO009586-5GEA 0 GBP
FLUO009586-5G FLUO009586-25G
2,4,6-Trimethylphenyl isothiocyanate
2,4,6-Trimethylphenyl isothiocyanate
Formula: C₁₀H₁₁NS
MW: 177.27 g/mol
Boiling Pt: 75 °C (0.3 mmHg)
Melting Pt: 64 °C
MDL Number: MFCD00022055
CAS Number: 6095-82-5
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR