2',5'-Dimethylacetophenone

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,5-Dimethylphenyl)ethanone, 1-(2,5-Dimethylphenyl)ethan-1-one

D4890.25 D4890.100
XXXXD4890.25EA 0 GBP
XXXXD4890.25 XXXXD4890.100
2',5'-Dimethylacetophenone
2',5'-Dimethylacetophenone
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1,0154 g/cm³ (0 °C)
MDL Number: MFCD00014988
CAS Number: 2142-73-6
EINECS: 218-399-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR