Thymol

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 6-Isopropyl-m-cresol2-Isopropyl-5-methylphenol4-Isopropyl-3-hydroxytoluene

ROTH5391.1EA 18.1 GBP
ROTH5391.1
Thymol
Thymol
Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 49…51 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002309
CAS Number: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8,III
Merck Index: 12,09540
https://uk.vwr.com/stibo/web/std.lang.all/78/44/2297844.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR