2-Ethyl-6-methylphenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E2265.10 E2265.50
XXXXE2265.10EA 0 GBP
XXXXE2265.10 XXXXE2265.50
2-Ethyl-6-methylphenyl isothiocyanate
2-Ethyl-6-methylphenyl isothiocyanate
Formula: C₁₀H₁₁NS
Boiling Pt: 88…91 °C (1013 hPa)
Density: 1,06 g/cm³ (20 °C)
MDL Number: MFCD00041068
CAS Number: 66609-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR