5-Fluoro-m-xylene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethylfluorobenzene, 1-Fluoro-3,5-dimethylbenzene, 5-Fluoro-1,3-dimethylbenzene

F1558-G.5 F1558-G.25
XXXXF1558-G.5EA 0 GBP
XXXXF1558-G.5 XXXXF1558-G.25
5-Fluoro-m-xylene
5-Fluoro-m-xylene
Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 145 °C (1013 hPa)
MDL Number: MFCD00052366
CAS Number: 461-97-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR