5-Bromo-2-fluoro-m-xylene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B2809.5 B2809.25
XXXXB2809.5EA 0 GBP
XXXXB2809.5 XXXXB2809.25
5-Bromo-2-fluoro-m-xylene
5-Bromo-2-fluoro-m-xylene
Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD01320701
CAS Number: 99725-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR