4,4'-Methylenedianiline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4,4’-Diaminodiphenylmethane4,4'-DiaminodiphenylmethaneBis(4-aminophenyl)methane4,4'-Methylenedianiline

SIAL32950-1KGEA 131 GBP
SIAL32950-1KG SIAL32950-250G SIAL32950-50G
4,4'-Methylenedianiline, Sigma-Aldrich®
4,4'-Methylenedianiline
Formula: C₁₃H₁₄N₂
MW: 198.27 g/mol
Boiling Pt: 398 °C (1013 hPa)
Melting Pt: 91…92 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007914
CAS Number: 101-77-9
EINECS: 202-974-4
UN: 2651
ADR: 6.1,III
Merck Index: 13,03000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR