4,4'-Methylenedianiline, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4,4’-Diaminodiphenylmethane
, 4,4'-Diaminodiphenylmethane
, Bis(4-aminophenyl)methane
, 4,4'-Methylenedianiline
M-Clarity™ quality level = MQ100
31640-250MGEA
73.4
GBP
31640-250MG
4,4'-Methylenedianiline, Supelco®
4,4'-Methylenedianiline
Formula:
C₁₃H₁₄N₂ MW: 198.27 g/mol Boiling Pt: 398 °C (1013 hPa) Melting Pt: 91…92 °C Density: 1.15 g/cm³ (20 °C) Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007914 CAS Number: 101-77-9 EINECS: 202-974-4 UN: 2651 ADR: 6.1,III Merck Index: 13,03000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...