Bromocresol green

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

ROTHT115.1EA 16.78 GBP
ROTHT115.1
Bromocresol green
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://uk.vwr.com/stibo/web/std.lang.all/13/36/2301336.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR