Butylbenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: n-Butylbenzene

M-Clarity™ quality level = MQ100

B90203-100ML B90203-25ML B90203-500ML
SIALB90203-100MLEA 103 GBP
SIALB90203-100ML SIALB90203-25ML SIALB90203-500ML
Butylbenzene, Sigma-Aldrich®
Butylbenzene
Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00009463
CAS Number: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3,III
Merck Index: 13,01548

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR