4-Toluenethiol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Mercapto-4-methylbenzene, 4-Methylbenzene-1-thiol, 4-Thiocresol, 4-Methylbenzenethiol, p-Toluenethiol, p-Thiocresol
M-Clarity™ quality level = MQ200
T28525-100G
T28525-500G
T28525-5G
SIALT28525-100GEA
122
GBP
SIALT28525-100G
SIALT28525-500G
SIALT28525-5G
4-Toluenethiol, Sigma-Aldrich®
4-Toluenethiol
Formula:
C₇H₈S MW: 124.21 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: 39…42 °C Density: 1.022 g/cm³ (51 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004851 CAS Number: 106-45-6 EINECS: 203-399-1 UN: 2811 ADR: 6.1,III Merck Index: 12,09461 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...