3-Toluenethiol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Thiocresol, 3-Methylbenzene-1-thiol, 3-Thiocresol, 3-Methylbenzenethiol, m-Toluenethiol, 1-Mercapto-3-methylbenzene, 3-Methylthiophenol

005338-5G 005338-25G 005338-100G
FLUO005338-5GEA 0 GBP
FLUO005338-5G FLUO005338-25G FLUO005338-100G
3-Toluenethiol
3-Toluenethiol
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 195 °C (1013 hPa)
Density: 1.06251 g/cm³ (0 °C)
MDL Number: MFCD00004843
CAS Number: 108-40-7
EINECS: 203-578-4
UN: 2810
ADR: 6.1,III
Merck Index: 12,09461

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR