4-tert-Butylphenol

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

506761.EA 61 GBP
506761.
4-tert-Butylphenol
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 95…98 °C
Density: 1.017 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 2430
ADR: 8,III
Merck Index: 13,01584

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR