3,5-Dinitrobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

184144-100G 184144-25G 184144-5G
SIAL184144-100GEA 1160 GBP
SIAL184144-100G SIAL184144-25G SIAL184144-5G
3,5-Dinitrobenzyl alcohol, Sigma-Aldrich®
3,5-Dinitrobenzyl alcohol
Formula: C₇H₆N₂O₅
MW: 198.14 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00007235
CAS Number: 71022-43-0
EINECS: 275-131-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR