3,4-Diaminotoluene 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Diamino-4-methylbenzene, Toluene-3,4-diamine

112431000 112435000
ACRO112431000EA 29.6 GBP
ACRO112431000 ACRO112435000
3,4-Diaminotoluene 97%
3,4-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 265 °C (760 mmHg)
Melting Pt: 83…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007728
CAS Number: 496-72-0
EINECS: 207-826-2

Order Now

Specification Test Results

Appearance Grey to brown Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 83 to 90 °C
HPLC ≥96.0 %
Absorption Ratio P-15/P+15 1.38 to 2.24
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR