3-Ethylaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

175498-10G 175498-50G
SIAL175498-10GEA 106 GBP
SIAL175498-10G SIAL175498-50G
3-Ethylaniline, Sigma-Aldrich®
3-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007818
CAS Number: 587-02-0
EINECS: 209-594-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR