Phenethylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: β-Phenylethylamine2-PhenylethylaminePEA

M-Clarity™ quality level = MQ100

SIAL241008-50MLEA 64.9 GBP
SIAL241008-50ML
Phenethylamine, Sigma-Aldrich®
Phenethylamine
Formula: C₆H₅CH₂CH₂NH₂
MW: 121.18 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.9639 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00008184
CAS Number: 64-04-0
EINECS: 200-574-4
UN: 2735
ADR: 8,I
Merck Index: 12,07371

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR