m-Tolyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001281-5G
001281-25G
001281-100G
FLUO001281-5GEA
0
GBP
FLUO001281-5G
FLUO001281-25G
FLUO001281-100G
m-Tolyl isothiocyanate
m-Tolyl isothiocyanate
Formula:
C₈H₇NS MW: 149.22 g/mol Boiling Pt: 243 °C (1013 hPa) Melting Pt: –20 °C Density: 1.103 g/cm³ (20 °C) |
MDL Number:
MFCD00004807 CAS Number: 621-30-7 EINECS: 210-677-6 UN: 2922 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...