2-Isopropylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008988-5G 008988-25G 008988-1G
FLUO008988-5GEA 0 GBP
FLUO008988-5G FLUO008988-25G FLUO008988-1G
2-Isopropylphenyl isothiocyanate
2-Isopropylphenyl isothiocyanate
Formula: C₁₀H₁₁NS
MW: 177.27 g/mol
Boiling Pt: 82 °C (1013 hPa)
Density: 1,05 g/cm³ (20 °C)
MDL Number: MFCD00022053
CAS Number: 36176-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR