o-Tolyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methylphenyl isocyanate, 1-Isocyanato-2-methylbenzene
M-Clarity™ quality level = MQ100
T40703-10G
T40703-50G
SIALT40703-10GEA
136
GBP
SIALT40703-10G
SIALT40703-50G
o-Tolyl isocyanate, Sigma-Aldrich®
o-Tolyl isocyanate
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 185 °C (1013 hPa) Density: 1.07 g/cm³ (20 °C) |
MDL Number:
MFCD00002009 CAS Number: 614-68-6 EINECS: 210-389-0 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...