3-(Trifluoromethyl)benzenesulphonyl chloride ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-(Trifluoromethyl)benzene-1-sulphonyl chloride

A14657.03 A14657.06 A14657.14
A14657.03EA 17.6 GBP
A14657.03 A14657.06 A14657.14
3-(Trifluoromethyl)benzenesulphonyl chloride ≥98%
3-(Trifluoromethyl)benzenesulphonyl chloride

Purity: 98% CAS No.: 777-44-6 Molecular Formula: C7H4ClF3O2S Formula Weight: 244.62 Flash Point: 110°(230°F) Refractive Index: 1.4870 Boiling Point: 79-80°/1mm Beilstein Reference: 2650774 UN No.: 3265 MDL No.: MFCD00014724

Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 79…80 °C (1 mmHg)
Density: 1.526
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014724
CAS Number: 777-44-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR