2,4-Difluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4-Difluorobenzyl Alcohol, (2,4-Difluorophenyl)methanol

001921-25G 001921-100G
FLUO001921-25GEA 0 GBP
FLUO001921-25G FLUO001921-100G
2,4-Difluorobenzyl alcohol
2,4-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 52 °C (1013 hPa)
Density: 1,29 g/cm³ (20 °C)
MDL Number: MFCD00009983
CAS Number: 56456-47-4
EINECS: 260-191-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR