2,3-Difluorobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

SIAL290262-5GEA 55.3 GBP
SIAL290262-5G
2,3-Difluorobenzyl alcohol, Sigma-Aldrich®
2,3-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00010312
CAS Number: 75853-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR