2,3-Difluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001862-5G
001862-25G
FLUO001862-5GEA
0
GBP
FLUO001862-5G
FLUO001862-25G
2,3-Difluorobenzyl alcohol
2,3-Difluorobenzyl alcohol
Formula:
C₇H₆F₂O MW: 144.12 g/mol Boiling Pt: 197 °C (1013 hPa) Melting Pt: 9 °C Density: 1,28 g/cm³ (20 °C) |
MDL Number:
MFCD00010312 CAS Number: 75853-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...