2,3,4,5,6-Pentafluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001352-10G
001352-25G
001352-100G
FLUO001352-10GEA
0
GBP
FLUO001352-10G
FLUO001352-25G
FLUO001352-100G
2,3,4,5,6-Pentafluorobenzyl alcohol
2,3,4,5,6-Pentafluorobenzyl alcohol
Formula:
C₇H₃F₅O Boiling Pt: 181 °C (1013 hPa) Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004602 CAS Number: 440-60-8 EINECS: 207-126-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...