α,α,α-Trifluoro-p-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A

001401-250G 001401-1KG
FLUO001401-250GEA 0 GBP
FLUO001401-250G FLUO001401-1KG
α,α,α-Trifluoro-p-toluidine
α,α,α-Trifluoro-p-toluidine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 3…8 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064396
CAS Number: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR