3-Fluoro-p-toluidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Fluoro-4-methylaniline, 4-Amino-2-fluorotoluene
M-Clarity™ quality level = MQ100
222666-25G
222666-5G
SIAL222666-25GEA
75.3
GBP
SIAL222666-25G
SIAL222666-5G
3-Fluoro-p-toluidine, Sigma-Aldrich®
3-Fluoro-p-toluidine
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 97 °C (15 mmHg) Melting Pt: 37 °C Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00007762 CAS Number: 452-77-7 EINECS: 207-212-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...