2-Fluoro-p-toluidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene
M-Clarity™ quality level = MQ100
337110-1G
337110-5G
SIAL337110-1GEA
43.5
GBP
SIAL337110-1G
SIAL337110-5G
2-Fluoro-p-toluidine, Sigma-Aldrich®
2-Fluoro-p-toluidine
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 190 °C (1013 hPa) Melting Pt: 3 °C Density: 1.1084 g/cm³ (18 °C) |
MDL Number:
MFCD00040975 CAS Number: 452-80-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...