2,4,5-Trifluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
007965-1G
007965-5G
FLUO007965-1GEA
0
GBP
FLUO007965-1G
FLUO007965-5G
2,4,5-Trifluorobenzylamine
2,4,5-Trifluorobenzylamine
Formula:
C₇H₆F₃N MW: 161.13 g/mol |
MDL Number:
MFCD00236316 CAS Number: 168644-93-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...