4-Fluoro-3-iodotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009207-10G
009207-25G
009207-5G
FLUO009207-10GEA
0
GBP
FLUO009207-10G
FLUO009207-25G
FLUO009207-5G
4-Fluoro-3-iodotoluene
4-Fluoro-3-iodotoluene
Formula:
C₇H₆FI MW: 236.03 g/mol |
MDL Number:
MFCD00079759 CAS Number: 452-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...