4-Nitro-α,α,α-trifluoro-m-toluic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
638714-1G
638714-5G
SIAL638714-1GEA
62.5
GBP
SIAL638714-1G
SIAL638714-5G
4-Nitro-α,α,α-trifluoro-m-toluic acid, Sigma-Aldrich®
4-Nitro-α,α,α-trifluoro-m-toluic acid
Formula:
C₈H₄F₃NO₄ MW: 235.12 g/mol |
MDL Number:
MFCD06200716 CAS Number: 320-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...