2',3',4',5',6'-Pentafluoroacetophenone ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L02063.03 L02063.06
L02063.03EA 22.6 GBP
L02063.03 L02063.06
2',3',4',5',6'-Pentafluoroacetophenone ≥97%
2',3',4',5',6'-Pentafluoroacetophenone

Purity: 97% CAS No.: 652-29-9 Molecular Formula: C8H3F5O Formula Weight: 210.10 Flash Point: 65°(149°F) Refractive Index: 1.4366 Boiling Point: 130-131° Beilstein Reference: 2053225 MDL No.: MFCD00000296

Formula: C₈H₃F₅O
MW: 210.1 g/mol
Boiling Pt: 130…131 °C
Density: 1.476
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000296
CAS Number: 652-29-9
EINECS: 211-487-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR