2',5'-Difluoropropiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,5-Difluorophenyl)propan-1-one

004872-5G 004872-25G 004872-1G
FLUO004872-5GEA 0 GBP
FLUO004872-5G FLUO004872-25G FLUO004872-1G
2',5'-Difluoropropiophenone
2',5'-Difluoropropiophenone
Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00015509
CAS Number: 29112-90-1
EINECS: 249-445-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR