3',5'-Difluoropropiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3,5-Difluorophenyl)propan-1-one

004875-5G 004875-25G
FLUO004875-5GEA 0 GBP
FLUO004875-5G FLUO004875-25G
3',5'-Difluoropropiophenone
3',5'-Difluoropropiophenone
Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00061142
CAS Number: 135306-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR