2',6'-Difluoropropiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,6-Difluorophenyl)propan-1-one

004873-5G 004873-25G
FLUO004873-5GEA 0 GBP
FLUO004873-5G FLUO004873-25G
2',6'-Difluoropropiophenone
2',6'-Difluoropropiophenone
Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00015506
CAS Number: 85068-31-1
EINECS: 285-293-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR