2',3'-Difluoropropiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,3-Difluorophenyl)propan-1-one

004871-5G 004871-10G
FLUO004871-5GEA 0 GBP
FLUO004871-5G FLUO004871-10G
2',3'-Difluoropropiophenone
2',3'-Difluoropropiophenone
Formula: C₉H₈F₂O
MW: 170.156 g/mol
MDL Number: MFCD00061141
CAS Number: 236754-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR