(2,3,5-Trifluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3,5-Trifluorobenzyl cyanide, 2,3,5-Trifluorophenylacetonitrile

007697-1G 007697-5G
FLUO007697-1GEA 0 GBP
FLUO007697-1G FLUO007697-5G
(2,3,5-Trifluorophenyl)acetonitrile
(2,3,5-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
MW: 171.12 g/mol
MDL Number: MFCD00083546
CAS Number: 243666-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR