α,α,α,2,3,5,6-Heptafluorotoluene ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B24592.06 B24592.14
B24592.06EA 58.7 GBP
B24592.06 B24592.14
α,α,α,2,3,5,6-Heptafluorotoluene ≥98%
α,α,α,2,3,5,6-Heptafluorotoluene

Purity: 98% CAS No.: 651-80-9 Molecular Formula: C7HF7 Formula Weight: 218.07 Flash Point: 32°(89°F) Refractive Index: 1.3790 Boiling Point: 111-112° Beilstein Reference: 1978201 UN No.: 1993 MDL No.: MFCD00012385

Formula: C₇HF₇
MW: 218.07 g/mol
Boiling Pt: 111…112 °C
Density: 1.601
Flash Pt: 32 °C (89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012385
CAS Number: 651-80-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR