α,α,α,3,4,5-Hexafluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007493-1G 007493-5G
FLUO007493-1GEA 0 GBP
FLUO007493-1G FLUO007493-5G
α,α,α,3,4,5-Hexafluorotoluene
α,α,α,3,4,5-Hexafluorotoluene
Formula: C₇H₂F₆
MW: 200.08 g/mol
MDL Number: MFCD00153280
CAS Number: 80172-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR