3-Nitro-p-toluic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methyl-3-nitrobenzoic acid
M-Clarity™ quality level = MQ100
151408-100G
151408-25G
SIAL151408-100GEA
61.5
GBP
SIAL151408-100G
SIAL151408-25G
3-Nitro-p-toluic acid, Sigma-Aldrich®
3-Nitro-p-toluic acid
Formula:
C₈H₇NO₄ MW: 181.15 g/mol Boiling Pt: 360 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00007174 CAS Number: 96-98-0 EINECS: 202-549-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...