2',6'-Dihydroxyacetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-(2,6-Dihydroxyphenyl)ethan-1-one2,6-Dihydroxyacetophenone1-(2,6-Dihydroxyphenyl)ethanone

M-Clarity™ quality level = MQ100

SIALD107808-10GEA 37.2 GBP
SIALD107808-10G
2',6'-Dihydroxyacetophenone, Sigma-Aldrich®
2',6'-Dihydroxyacetophenone
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002270
CAS Number: 699-83-2
EINECS: 211-833-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR