2',6'-Dihydroxyacetophenone
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(2,6-Dihydroxyphenyl)ethan-1-one
, 2,6-Dihydroxyacetophenone
, 1-(2,6-Dihydroxyphenyl)ethanone
PROOMM0039.03EA
857
GBP
PROOMM0039.03
2',6'-Dihydroxyacetophenone
2',6'-Dihydroxyacetophenone
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002270 CAS Number: 699-83-2 EINECS: 211-833-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...