3,5-Dichlorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013039-5G
013039-25G
FLUO013039-5GEA
0
GBP
FLUO013039-5G
FLUO013039-25G
3,5-Dichlorobenzyl alcohol
3,5-Dichlorobenzyl alcohol
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Boiling Pt: 264 °C (1013 hPa) Melting Pt: 82 °C |
MDL Number:
MFCD00004634 CAS Number: 60211-57-6 EINECS: 262-109-1 UN: 2250 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...