2,6-Dichlorobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,6-Dichlorophenyl)methanol

M-Clarity™ quality level = MQ100

100293-10G 100293-25G
SIAL100293-10GEA 52.2 GBP
SIAL100293-10G SIAL100293-25G
2,6-Dichlorobenzyl alcohol, Sigma-Aldrich®
2,6-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96…98 °C
MDL Number: MFCD00004609
CAS Number: 15258-73-8
EINECS: 239-300-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR