2-Chlorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (2-Chlorophenyl)methanamine

008868-10G 008868-25G
FLUO008868-10GEA 0 GBP
FLUO008868-10G FLUO008868-25G
2-Chlorobenzylamine
2-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 103 °C (1013 hPa)
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008108
CAS Number: 89-97-4
EINECS: 201-955-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR