4-Chloro-2-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-chloro-α,α,α-trifluorotoluene, 2-Amino-5-chloro-α,α,α-trifluortoluene, 2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine

002680-100G 002680-500G
FLUO002680-100GEA 0 GBP
FLUO002680-100G FLUO002680-500G
4-Chloro-2-(trifluoromethyl)aniline
4-Chloro-2-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007841
CAS Number: 445-03-4
EINECS: 207-151-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR