4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008778-5G
008778-1G
FLUO008778-5GEA
0
GBP
FLUO008778-5G
FLUO008778-1G
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Formula:
C₈H₃ClF₃NS MW: 237.63 g/mol Boiling Pt: 270 °C (1013 hPa) Melting Pt: 34…36 °C Density: 1,44 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00041090 CAS Number: 23163-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...