α-Chloro-2,4-difluorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

021891-5G 021891-25G
FLUO021891-5GEA 0 GBP
FLUO021891-5G FLUO021891-25G
α-Chloro-2,4-difluorotoulene
α-Chloro-2,4-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
MDL Number: MFCD01090988
CAS Number: 452-07-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR