4-Bromo-3-methylanisole

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

B2944-D.25 B2944-D.100
XXXXB2944-D.25EA 0 GBP
XXXXB2944-D.25 XXXXB2944-D.100
4-Bromo-3-methylanisole
4-Bromo-3-methylanisole
Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR